Learn More
HLA-E Mouse anti-Human, Janelia Fluor 646, Clone: 3D12HLA-E, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP143124JF646
Catalog No. NBP43124646
Add to cart
Description
Description
HLA-E Monoclonal antibody specifically detects HLA-E in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
HLA-E
3D12HLA-E
Flow Cytometry
Purified
HLA-E
The immunogen for this antibody was HLAE.
Protein A or G purified
Adaptive Immunity, Asthma, Diabetes Research, Immunology, Major Histocompatibility Complexes
Primary
3133
Flow Cytometry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1
Mouse
IgG1 Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Human