Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA F Mouse anti-Human, DyLight™ 350, Clone: 3D11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP276802UV

Catalog No. NBP276802UV


Add to cart

Description

Description

HLA F Monoclonal antibody specifically detects HLA F in Human samples. It is validated for Western Blot,Flow Cytometry.
Specifications

Specifications

HLA F
3D11
Western Blot, Flow Cytometry
alpha chain F, HLA class I molecule, HLA F antigen, HLA-5.4, HLAFHLA-CDA12, Leukocyte antigen F, major histocompatibility complex, class I, F, MHC class I antigen F, MHC class Ib antigen
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Western Blot
DyLight™ 350
50 mM sodium borate with 0.05% sodium azide
HLA-F
Inclusion body-derived HLA-F heavy chain
0.1 mL
Primary
3134
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit