Learn More
HLA-G Mouse anti-Human, BUV395, Clone: 87G, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 751661
Catalog No. BDB751661
Add to cart
Description
Description
HLA-G Monoclonal antibody specifically detects HLA-G in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
HLA-G
Flow Cytometry
0.2 mg/mL
Hu HLA-G BUV395 87G 50 μg
Mouse
IgG2a, κ
RUO
Primary
HLA-G
Human
87G
Brilliant Ultraviolet 395
BUV395
HLA-G-transfected Cells
1 x 50 μg
Refrigerate (2 to 8°C)
Monoclonal
Human