Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-G Mouse anti-Human, FITC, Clone: MEM-G/9, Invitrogen™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen A15741

Catalog No. 50-113-7089


Add to cart

Description

Description

HLA-G belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. HLA-G is expressed on fetal derived placental cells. The heavy chain is approximately 45 kDa and its gene contains 8 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domain, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exon 6 encodes the cytoplasmic tail.
Specifications

Specifications

HLA-G
Monoclonal
1 mg/mL
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
P17693
DADB-15K14.8, MHC-G, HLA G antigen, HLA class I histocompatibility antigen, alpha chain G, HLA class I molecule, HLA-G histocompatibility antigen, class I, G, MHC class I antigen G, b2 microglobulin
Mouse
IgG1
100 μg
4° C
Primary
3135
Flow Cytometry
MEM-G/9
FITC
HLA-G
Liquid
HLA-G
human HLA-G
Purified
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit