Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA C Mouse anti-Human, Mouse, Janelia Fluor 549, Clone: DT-9, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250419JF549
Description
HLA C Monoclonal specifically detects HLA C in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
HLAC | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HLA-C | |
Purified tamarin MHC class I molecules (using anti-class I antibody BB7.7) | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry | |
DT-9 | |
Flow Cytometry | |
C alpha chain, D6S204, FLJ27082, HLA class I histocompatibility antigen, Cw-1 alpha chain, HLAC, HLA-Cw, HLA-JY3, HLC-C, human leukocyte antigen-C alpha chain, leucocyte antigen C, leucocyte antigen Cw, leukocyte antigen-C alpha chain, major histocompatibility antigen HLA-C, major histocompatibility complex, class I, C, MHC class I antigen Cw*1, MHC class I antigen Cw*12, MHC class I antigen Cw*14, MHC class I antigen Cw*15, MHC class I antigen Cw*16, MHC class I antigen Cw*17, MHC class I antigen Cw*2, MHC class I antigen Cw*3, MHC class I antigen Cw*4, MHC class I antigen Cw*5, MHC class I antigen Cw*6, MHC class I antigen Cw*7, MHC class I antigen Cw*8, MHC class I antigen heavy chain HLA-C, PSORS1 | |
Mouse | |
Protein G purified | |
Adaptive Immunity, Immunology | |
3107 | |
Store at 4°C in the dark. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title