Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HM74A/PUMA-G/GPR109A/NIACR1 Antibody (MM0329-6S24), Alexa Fluor™ 594, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212358AF594
Description
HM74A/PUMA-G/GPR109A/NIACR1 Monoclonal specifically detects HM74A/PUMA-G/GPR109A/NIACR1 in Human samples. It is validated for Flow Cytometry.Specifications
HM74A/PUMA-G/GPR109A/NIACR1 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
HCAR2 | |
Human recombinant HM74A/PUMA-G/GPR109A/NIACR1 | |
Primary | |
Human | |
IgG2 |
Western Blot, Flow Cytometry, Immunohistochemistry | |
MM0329-6S24 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
GPR109A, HCA2, hydroxycarboxylic acid receptor 2, NIACR1 | |
Rat | |
0.1 mL | |
338442 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction