Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMBS Rabbit anti-Human, Clone: SC65-07, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP267637

Catalog No. NBP267637


Add to cart

Description

Description

HMBS Monoclonal antibody specifically detects HMBS in Human samples. It is validated for Western Blot.
Specifications

Specifications

HMBS
SC65-07
Unconjugated
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide
hydroxymethylbilane synthasePre-uroporphyrinogen synthase, PBGDPBG-D, PORC, porphobilinogen deaminase, porphyria, acute; Chester type, UPSEC 2.5.1.61, uroporphyrinogen I synthase, uroporphyrinogen I synthetase
Rabbit
IgG
100μL
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse, Rat
Western Blot
1 mg/mL
Western Blot 1:1000-1:2000
Purified
HMBS
Full length protein
Protein A purified
Diabetes Research
Primary
3145
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit