Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMBS Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$152.22 - $436.00
Specifications
Antigen | HMBS |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15662720
|
Novus Biologicals
NBP15662720UL |
20 μL |
Each for $152.22
|
|
NBP156627
|
Novus Biologicals
NBP156627 |
100 μL |
Each for $436.00
|
|
Description
HMBS Polyclonal specifically detects HMBS in Human samples. It is validated for Western Blot.Specifications
HMBS | |
Polyclonal | |
Rabbit | |
Diabetes Research | |
P08397 | |
3145 | |
Synthetic peptides corresponding to HMBS(hydroxymethylbilane synthase) The peptide sequence was selected from the middle region of HMBS. Peptide sequence SSLRRAAQLQRKFPHLEFRSIRGNLNTRLRKLDEQQEFSAIILATAGLQR. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
hydroxymethylbilane synthasePre-uroporphyrinogen synthase, PBGDPBG-D, PORC, porphobilinogen deaminase, porphyria, acute; Chester type, UPSEC 2.5.1.61, uroporphyrinogen I synthase, uroporphyrinogen I synthetase | |
HMBS | |
IgG | |
Affinity Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title