Learn More
HMCN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 188371AP
Catalog No. 50-173-2253
Add to cart
Description
Description
This antibody raised against a synthesized peptide corresponding to 960-916 amino acids of human HMCN1 can recognize all three isoforms of HMCN1.
Hemicentin-1 (HMCN1), also known as fibulin-6 (FIBL-6), is an extracellular protein of the immunoglobulin superfamily. HMCN1 consists of a von Willebrand A domain, tandem immunoglobulin modules, tandem EGF modules, and a fibulin carboxy-terminal module. Mutations of HMCN1 gene may be associated with age-related macular degeneration. Three alternative transcripts encoding different HMCN1 isoforms have been described.
Specifications
Specifications
HMCN1
0.71 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
D3YXG0, Q96RW7
ARMD1, FBLN6, FIBL 6, FIBL6, Fibulin 6, hemicentin 1, HMCN1
Rabbit
IgG
150 μL
-20°C
Primary
289094, 545370, 83872
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
HMCN1
Liquid
Hmcn1
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat