Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMG-1 Rabbit anti-Human, Unconjugated, Polyclonal, Active Motif™ Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

$135.30 - $534.00

Specifications

Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Antigen HMG-1
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-4049 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
39552
10 μL
Each for $135.30
Add to cart
 
50-199-4048 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
39551
200 μL
Each for $534.00
Add to cart
 
Description

Description

BHMG1 gene ontology annotations related to this gene include DNA binding; protein dimerization activity.
Specifications

Specifications

Polyclonal
Primary
HMG-1
Unconjugated
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit