Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMG-CoA Reductase/HMGCR Antibody (JF0981), Novus Biologicals™
SDP

Rabbit Monoclonal Antibody has been used in 1 publication

Supplier:  Novus Biologicals NBP266888

Catalog No. NBP266888


Add to Cart

Description

Description

HMG-CoA Reductase/HMGCR Monoclonal specifically detects HMG-CoA Reductase/HMGCR in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunoprecipitation.
Specifications

Specifications

HMG-CoAReductase/HMGCR
Monoclonal
1 mg/mL
Western Blot 1:500-1:2000, Simple Western, Immunoprecipitation, Immunohistochemistry-Paraffin 1:100
3-hydroxy-3-methylglutaryl CoA reductase (NADPH), 3-hydroxy-3-methylglutaryl-CoA reductase, 3-hydroxy-3-methylglutaryl-Coenzyme A reductase, EC 1.1.1, EC 1.1.1.34, HMG-CoA reductase, hydroxymethylglutaryl-CoA reductase, LDLCQ3
Rabbit
Protein A purified
Primary
Human, Mouse, Rat
Purified
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
JF0981
Unconjugated
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
HMGCR
Synthetic peptide within Human HMG-CoA Reductase/HMGCR aa 438-489/ 888. (SwissProt: P04035 Human; SwissProt: Q01237 Mouse; SwissProt: P51639 Rat)
100 μL
3156
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit