Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HMG20A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587308
Description
Purity is > 95% by SDS-PAGE.
Plays a role in neuronal differentiation as chromatin-associated protein. Acts as inhibitor of HMG20B. Overcomes the repressive effects of the neuronal silencer REST and induces the activation of neuronal-specific genes. Involved in the recruitment of the histone methyltransferase KMT2A/MLL1 and consequent increased methylation of histone H3 lysine 4.
Specifications
HMG20A | |
Polyclonal | |
Unconjugated | |
HMG20A | |
1200004E06Rik; 5730490E10Rik; FLJ10739; high mobility group 20A; high mobility group protein 20A; HMG box domain containing 1; HMG box-containing protein 20A; HMG domain-containing protein 1; HMG domain-containing protein HMGX1; HMG20A; HMGX1; HMGXB1; Ibraf; Inhibitor of BRAF35; RGD1564760 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10363, 66867 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry, Western Blot, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9DC33, Q9NP66 | |
HMG20A | |
Recombinant full length Human HMG20A. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction