Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMG2L1 Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI1H10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271538AF647

Catalog No. N271538A647


Add to cart

Description

Description

HMG2L1 Monoclonal antibody specifically detects HMG2L1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

HMG2L1
OTI1H10
Western Blot
Affinity Purified, Conjugated
HMGXB4
Recombinant protein fragment corresponding to amino acids 1-348 of human HMG2L1 (NP_005478) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
High mobility group protein 2-like 1, high-mobility group protein 2-like 1, HMG box domain containing 4, HMG domain-containing protein 4, HMG2L1HMG box-containing protein 4, HMGBCG, Protein HMGBCG, THC211630
Mouse
IgG1
0.1 mL
Primary
10042.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit