Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMGA2 Mouse anti-Human, Mouse, Rat, Alexa Fluor™ 350, Clone: 763, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243640AF350

Catalog No. NP243640A30


Add to cart

Description

Description

HMGA2 Monoclonal antibody specifically detects HMGA2 in Human,Mouse,Rat samples. It is validated for Western Blot,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin).
Specifications

Specifications

HMGA2
763
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
BABLHMGI-C, high mobility group AT-hook 2, High mobility group AT-hook protein 2, high mobility group protein HMGI-C, high-mobility group (nonhistone chromosomal) protein isoform I-C, High-mobility group protein HMGI-C, HMGICSTQTL9, LIPO
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
HMGA2
Recombinant protein encompassing a sequence within the center region of human HMGA2. The exact sequence is proprietary.
0.1 mL
Primary
8091
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit