Learn More
HMGB1/HMG-1 Mouse anti-Human, Mouse, Alexa Fluor™ 350, Clone: 19N15F4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP227401AF350
Catalog No. NP227401A30
Add to cart
Description
Description
HMGB1/HMG-1 Monoclonal antibody specifically detects HMGB1/HMG-1 in Human,Mouse samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
HMGB1/HMG-1
19N15F4
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
This antibody recognizes both the box A domain (amino acids 9-79) and full length HMGB1 protein.
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
HMGB1
Full-length recombinant human HMGB1 was used as the immunogen for the antibody.
0.1 mL
Primary
3146
Human, Mouse