Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMGB1 Mouse anti-Human, Clone: IMG19N10B7, Abnova™

Mouse monoclonal antibody raised against human HMGB1.

Manufacturer:  Abnova Corporation MAB12722

Catalog No. 89-934-281


Add to cart

Description

Description

Specifications

Specifications

HMGB1
IMG19N10B7
Mouse monoclonal antibody raised against human HMGB1.
In PBS.
DKFZp686A04236/HMG1/HMG3/SBP-1
Mouse
IgG2b, kappa
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
HMGB1
Recombinant protein corresponding to full length human HMGB1.
Protein G purification
RUO
Primary
3146
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit