Learn More
HMGB1 Rabbit anti-Human,Mouse,Rat, KHL, Clone: 3K6, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB100225UL
 View more versions of this product
Catalog No. ZRB100225MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
HMGB1 Monoclonal antibody specifically detects HMGB1 in Human,Mouse,Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications
Specifications
HMGB1
3K6
KHL
Purified
HMGB1, HMG1
KLH-conjugated linear peptide corresponding to 13 amino acids from the C-terminal half of human High mobility group protein B1.
∼30 kDa observed; 24.89 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
30 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
NP_002119.1
High mobility group protein B1;High mobility group protein 1;HMG-1
Rabbit
IgG
Protein A Purified
Epigenetics and Nuclear Function
Primary
Human, Mouse, Rat