Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ HMGCL Recombinant Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$280.50 - $1004.00

Specifications

Formulation PBS (pH 7.4) containing 20% glycerol, 1mM DTT. containingno Preservative
Gene Alias 3-hydroxy-3-methylglutaryl-CoA lyase, 3-hydroxymethyl-3-methylglutaryl-CoA lyase, EC 4.1.3.4, HLmitochondrial
Gene ID (Entrez) 3155
Gene Symbol HMGCL
Name Recombinant HMGCL Protein
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB60212250U View Documents Novus Biologicals™
NBP260212250UG
250μg Each for $1,004.00
Add to cart
 
NB6021250UG View Documents Novus Biologicals™
NBP26021250UG
50μg Each for $280.50
Add to cart
 
Description

Description

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 28-325 of Human HMGCL The Recombinant Human HMGCL Protein is derived from Baculovirus. The Recombinant Human HMGCL Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

PBS (pH 7.4) containing 20% glycerol, 1mM DTT. containingno Preservative
3155
Recombinant HMGCL Protein
Baculovirus
1mg/mL
SDS-Page
3-hydroxy-3-methylglutaryl-CoA lyase, 3-hydroxymethyl-3-methylglutaryl-CoA lyase, EC 4.1.3.4, HLmitochondrial
HMGCL
Recombinant Protein
RUO
< 1.0 EU per 1ug of protein (determined by LAL method)

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit