Learn More
BioVision IncSupplier Diversity Partner HMG-CoA Reductase Activity/Inhibitor Screening Kit (Colorimetric): Simple fluorometric assay to measure activity and screen inhibitors/activators of HMGR. Includes Inhibitor Control, Atorvastatin. 100 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K588100
HMG-CoA reductase (3-hydroxy-3-methyl-glutaryl-CoA reductase or HMGR) (EC 1.1.1.34) is the rate-controlling enzyme of the mevalonate pathway, the metabolic pathway that produces cholesterol from acetyl-CoA. In an NADPH-dependent reaction, HMG-CoA reductase reduces HMG-CoA to generate mevalonate and CoA. The enzyme is target of a group of cholesterol-lowering drugs known as statins. Inhibition of HMG-CoA reductase induces expression of LDL receptors in the liver, which lowers plasma concentration of cholesterol. BioVision’s HMG-CoA Reductase Activity Assay Kit measures utilization of NADPH, which can be measured by the decrease of absorbance (OD 340 nm). The kit is suitable for measuring activity of purified enzyme and screening inhibitors/activators of HMG-CoA reductase. The limit of detection is below 0.05 mU. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9784