Learn More
Abnova™ HMGCS2 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00003158T01
Catalog No. 89-947-641
Add to cart
Specifications
Specifications
Western Blot
293T
NM_005518.2
pCMV-HMGCS2 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
HMGCS2 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 55.99kDa
Transient overexpression cell lysate was tested with anti-HMGCS2 antibody (Mfr. No. H00003158-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)