Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HMGXB4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5139948
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: YQVFCKEYRV TIVADHPGID FGELSKKLAE VWKQLPEKDK LIWKQKAQYL QHKQNKAEAT TVKRKASSSE GSMKVKASSV GVLSPQKKSP PT.
High mobility group (HMG) proteins are nonhistone chromosomal proteins.
Specifications
HMGXB4 | |
Polyclonal | |
Unconjugated | |
HMGXB4 | |
high mobility group protein 2-like 1; high-mobility group protein 2-like 1; HMG box domain containing 4; HMG box-containing protein 4; HMG domain-containing protein 4; HMG2L1; HMGBCG; HMG-box containing 4; HMGXB4; Protein HMGBCG; Protein THC211630; THC211630 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10042 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UGU5 | |
HMGXB4 | |
Recombinant protein corresponding to Human HMGXB4. Recombinant protein control fragment (Product #RP-109581). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction