Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMHA1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549605

Catalog No. PA549605

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

GTPase activator for the Rho-type GTPases.
Specifications

Specifications

HMHA1
Polyclonal
PBS with 0.09% sodium azide
Q92619
Minor histocompatibility protein HA-1, Minor histocompatibility antigen HA-1, mHag HA-1, HMHA1, KIAA0223
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23526
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ARHGAP45
Liquid
ARHGAP45
KLH conjugated synthetic peptide between 877-903 amino acids from the Central region of human HMHA1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit