Learn More
HMHA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 148321AP
Catalog No. 50-172-9762
Add to cart
Description
Description
HMHA1 (ARHGAP45) Contains a GTPase activator for the Rho-type GTPases (RhoGAP) domain that would be able to negatively regulate the actin cytoskeleton as well as cell spreading. However, also contains N-terminally a BAR-domin which is able to play an autoinhibitory effect on this RhoGAP activity.
Specifications
Specifications
HMHA1
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3TBD2, Q92619
HA 1, HLA HA1, HMHA1, KIAA0223, mHag HA 1
Rabbit
IgG
150 μL
-20°C
Primary
23526, 314618, 70719
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ARHGAP45
Liquid
Arhgap45
HMHA1 Fusion Protein Ag6636
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat