Learn More
Brady™ HMIG Self-Laminating Labels
Include space to write in appropriate chemical ratings. Brady™ HMIG Self-Laminating Labels are made of polyester w/polyester laminate cover.
Manufacturer:  Brady™ 60333
 View more versions of this product
Catalog No. 19-110-410
  Check Availability
Add to cart
Specifications
Description
  • Include space to write in the appropriate chemical ratings as well as personal protective equipment index
  • Rectangular shape
  • Black, red, blue, and yellow on white
Description & Specifications
Specifications
Acrylic,Permanent
HEALTH FLAMMABILITY REACTIVITY PROTECTIVE EQUIPMENT
W x H: 5 x 3 in.
Polyester,Polyester Laminate
25/pk
Black,Blue,Red,White,Yellow
Rectangle
HMIG Color Bar
HMIG Self-Laminating Label