Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heme oxygenase 2 Mouse anti-Human, Clone: OTI1C2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525747

Catalog No. PIMA525747


Add to cart

Description

Description

Heme oxygenase, an essential enzyme in heme catabolism, cleaves heme to form biliverdin, which is subsequently converted to bilirubin by biliverdin reductase, and carbon monoxide, a putative neurotransmitter. Heme oxygenase activity is induced by its substrate heme and by various nonheme substances. Heme oxygenase occurs as 2 isozymes, an inducible heme oxygenase-1 and a constitutive heme oxygenase-2. HMOX1 and HMOX2 belong to the heme oxygenase family. Alternative splice variants encoding the same protein have been identified at this locus.
Specifications

Specifications

Heme oxygenase 2
Monoclonal
0.74 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P30519
HMOX2; heme oxygenase (decycling) 2; HO-2; heme oxygenase (decyclizing) 2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3163
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI1C2
Unconjugated
HMOX2
Liquid
HMOX2
Full length human recombit protein of HMOX2 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit