Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HNF-3 alpha/FoxA1 Mouse anti-Human, Rat (Negative), PE/Cy5.5, Clone: FOXA1/1514, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254575PECY55

Catalog No. N254575PEC5


Add to cart

Description

Description

HNF-3 alpha/FoxA1 Monoclonal antibody specifically detects HNF-3 alpha/FoxA1 in Human, Rat (Negative) samples. It is validated for Flow Cytometry.
Specifications

Specifications

HNF-3 alpha/FoxA1
FOXA1/1514
Flow Cytometry
Conjugated, Purified
FOXA1
Recombinant human FOXA1 protein fragment (aa372-472) (exact sequence is proprietary)
Protein A or G purified
Breast Cancer, Cancer, Stem Cell Markers
Primary
3169.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
forkhead box A1, Forkhead box protein A1, hepatocyte nuclear factor 3, alpha, hepatocyte nuclear factor 3-alpha, HNF-3A, HNF-3-alpha, HNF3ATCF-3A, MGC33105, TCF3A, Transcription factor 3A
Mouse
IgG2b Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Rat (Negative)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit