Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HNF-3 beta/FoxA2 Mouse anti-Human, PE-Cyanine7, Clone: OTI3C10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270904PECY7

Catalog No. N270904PEC7


Add to cart

Description

Description

HNF-3 beta/FoxA2 Monoclonal antibody specifically detects HNF-3 beta/FoxA2 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

HNF-3 beta/FoxA2
OTI3C10
Flow Cytometry
Affinity Purified, Conjugated
FOXA2
Recombinant protein expressed in E.coli corresponding to amino acids 300-458 of human HNF-3 beta/FoxA2.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
forkhead box A2, Forkhead box protein A2, hepatic nuclear factor-3-beta, hepatocyte nuclear factor 3, beta, hepatocyte nuclear factor 3-beta, HNF-3B, HNF-3-beta, HNF3BTCF-3B, MGC19807, TCF3B, Transcription factor 3B
Mouse
IgG1
0.1 mL
Primary
3170.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit