Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HNF-6/ONECUT1 Mouse anti-Human, Alexa Fluor™ 700, Clone: OTI4F12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270906AF700

Catalog No. N270906A700


Add to cart

Description

Description

HNF-6/ONECUT1 Monoclonal antibody specifically detects HNF-6/ONECUT1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

HNF-6/ONECUT1
OTI4F12
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
ONECUT1
Recombinant protein fragment corresponding to amino acids 85-198 of human HNF-6/ONECUT1 (NP_004489) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
HNF6Ahepatocyte nuclear factor 6, alpha, HNF6hepatocyte nuclear factor 6, HNF-6One cut domain family member 1, one cut homeobox 1one cut domain, family member 1
Mouse
IgG1
0.1 mL
Primary
3175.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit