Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HNF-6/ONECUT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP19160720UL
Description
HNF-6/ONECUT1 Polyclonal specifically detects HNF-6/ONECUT1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
HNF-6/ONECUT1 | |
Polyclonal | |
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
NP_032288 | |
ONECUT1 | |
Synthetic peptide directed towards ONECUT1. Peptide sequence CKRKEQEHGKDRGNTPKKPRLVFTDVQRRTLHAIFKENKRPSKELQITIS. | |
Affinity Purified | |
RUO | |
3175 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
Unconjugated | |
PBS & 2% Sucrose. with No Preservative | |
HNF6Ahepatocyte nuclear factor 6, alpha, HNF6hepatocyte nuclear factor 6, HNF-6One cut domain family member 1, one cut homeobox 1one cut domain, family member 1 | |
Rabbit | |
51 kDa | |
20 μL | |
Primary | |
Mouse | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction