Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HNF1, Mouse anti-Human, DyLight 650, Clone: HNF1A/2087, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279911C
Description
HNF1 Monoclonal antibody specifically detects HNF1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
HNF1 | |
HNF1A/2087 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Conjugated, Purified | |
HNF1A | |
Recombinant fragment (around aa 214-339) of human HNF1 protein (exact sequence is proprietary) | |
Protein A or G purified | |
RUO | |
Store at 4°C in the dark. | |
Monoclonal |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HNF1 homeobox A, IDDM20, LFB1transcription factor 1, hepatic; LF-B1, hepatic nuclear factor (HNF1), albuminproximal factor, MODY3, TCF1hepatic, Transcription factor 1 | |
Mouse | |
IgG1 Kappa | |
0.1 mL | |
Transcription Factors and Regulators | |
Primary | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title