Learn More
Aviva Systems Biology Corp HNF1A Antibody - N-terminal region (ARP51375_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP51375_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HNF1A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HNF1A. Alternate names: HNF1, LFB1, MODY3, TCF1, TCF-1, HNF-1A, IDDM20ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-60563