Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550531

Catalog No. PA550531


Add to cart

Description

Description

The antibody detects endogenous levels of total HNF1B protein.

This gene encodes a member of the homeodomain-containing superfamily of transcription factors. The protein binds to DNA as either a homodimer, or a heterodimer with the related protein hepatocyte nuclear factor 1-alpha. The gene has been shown to function in nephron development, and regulates development of the embryonic pancreas. Mutations in this gene result in renal cysts and diabetes syndrome and noninsulin-dependent diabetes mellitus, and expression of this gene is altered in some types of cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

TCF2
Polyclonal
Unconjugated
HNF1B
Liquid
HNF1B
Synthetic peptide corresponding to a region derived from internal residues of human HNF1 homeobox B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin)
2.6 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
P35680, P27889, P23899
FJHN, HNF2, LFB3, TCF2, HPC11, LF-B3, MODY5, TCF-2, VHNF1, HNF-1B, HNF1beta
Rabbit
IgG
100 μL
-20°C
Primary
21410, 25640, 6928
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit