Learn More
Alfa Aesar™ HNHA
Manufacturer:  Alfa Aesar™ J64917MA
Catalog No. AAJ64917MA
Order in Bulk
Request bulk or custom formats
Add to cart
Description
Description
A cell-permeable hydroxyamide-based SAHA analog that potently inhibits histone deacetylase (HDAC) activity.
Specifications
Specifications
10mg
Crystalline Solid
926908-04-5
303.42g/mol
Light sensitive
Crystalline solid
2° to 8°C
A cell-permeable hydroxyamide-based SAHA analog that potently inhibits histone deacetylase (HDAC) activity
C17H21NO2S
Histone Deacetylase Inhibitor VI
Signal Transduction Reagent
Safety and Handling
Safety and Handling
Recommended Storage : Keep cold