Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP F Mouse anti-Human, PE/Cy5.5, Clone: OTI5F5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270907PECY55

Catalog No. N270907PEC5


Add to cart

Description

Description

hnRNP F Monoclonal antibody specifically detects hnRNP F in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

hnRNP F
OTI5F5
Flow Cytometry
Affinity Purified, Conjugated
HNRNPF
Full length recombinant protein of human hnRNP F (NP_004957) produced in HEK293T cells.
Immunogen affinity purified
DNA Repair, DNA replication Transcription Translation and Splicing
Primary
3185.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
heterogeneous nuclear ribonucleoprotein F, hnRNP F, HNRPFHnRNP F protein, mcs94-1, MGC110997, Nucleolin-like protein mcs94-1, OK/SW-cl.23
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit