Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP H Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen hnRNP H
Applications Immunofluorescence
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095520
View Documents
Novus Biologicals
NBP303544100UL
100 μg
Each for $320.71
Add to cart
 
NB095521
View Documents
Novus Biologicals
NBP30354420UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

hnRNP H Polyclonal antibody specifically detects hnRNP H in Human samples. It is validated for Immunofluorescence
Specifications

Specifications

hnRNP H
Unconjugated
Rabbit
Cellular Markers, Translation Control
PBS with 50% glycerol, pH7.3.
3187
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence
Purified
RUO
Human
DKFZp686A15170, heterogeneous nuclear ribonucleoprotein H, heterogeneous nuclear ribonucleoprotein H1 (H), hnRNP H, HNRPH, HNRPH1hnRNPH
Recombinant fusion protein containing a sequence corresponding to amino acids 191-449 of human hnRNP H (NP_005511.1).
Polyclonal
Affinity purified
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit