Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP L Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen hnRNP L
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB092603
View Documents
Novus Biologicals
NBP303640100UL
100 μg Each for $320.71
Add to cart
 
NB092604
View Documents
Novus Biologicals
NBP30364020UL
20 μg Each for $138.37
Add to cart
 
Description

Description

hnRNP L Polyclonal antibody specifically detects hnRNP L in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications

Specifications

hnRNP L
Unconjugated
Rabbit
Cancer, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers
PBS with 50% glycerol, pH7.3.
3191
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence, Immunohistochemistry, Western Blot
Purified
RUO
Human, Mouse, Rat
FLJ35509, heterogeneous nuclear ribonucleoprotein L, hnRNP L, hnRNP-L, HNRPL
Recombinant fusion protein containing a sequence corresponding to amino acids 282-589 of human hnRNP L (NP_001524.2).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit