Learn More
hnRNP UL1 Rat anti-Human, Clone: EAP 4A11, eBioscience™ Invitrogen™
Rat Monoclonal Antibody
Manufacturer: LIFE TECHNOLOGIES 14980695

Description
This EAP 4A11 monoclonal antibody recognizes human hnRNP UL1. EAP 4A11 detects a ∽95 kDa band corresponding to hnRNP UL1 by Western Blot and stains hnRNP UL1 in the nucleus in Immunocytochemistry. Applications Reported: This EAP 4A11 antibody has been reported for use in western blot and immunocytochemistry/immunofluorescence. Applications Tested: This EAP 4A11 antibody has been tested by western blot of human cell lysates and immunocytochemistry/immunofluorescence of human cell lines. This may be used at less than or equal to 4.0 μg/ml for western blot and less than or equal to 2.5 μg/mL for immunocytochemistry/immunofluorescence.
hnRNP UL1 Monoclonal antibody specifically detects hnRNP UL1 in Human samples. It is validated for Immunocytochemistry, Immunofluorescence, Western BlotSpecifications
hnRNP UL1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q9BUJ2 | |
Rat | |
10 mg | |
Primary | |
Human | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
EAP 4A11 | |
Unconjugated | |
Hnrnpul1 | |
Adenovirus early region 1B-associated protein 5; E130317O14Rik; E1B 55kDa associated protein 5; E1B-55 kDa-associated protein 5; E1BAP5; E1B-AP5; EIB-55kDa associated protein 5; heterogeneous nuclear ribonucleoprotein U like 1; heterogeneous nuclear ribonucleoprotein U-like 1; heterogeneous nuclear ribonucleoprotein U-like protein 1; Hnrnpul; Hnrnpul1; HNRPUL1 | |
Affinity chromatography | |
RUO | |
11100 | |
Antibody | |
IgG2b κ |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.