Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HNRNPA0 Mouse anti-Human, Clone: 10F1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP19129020UL

Catalog No. NBP1929020


Add to cart

Description

Description

HNRNPA0 Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

HNRNPA0
10F1
Western Blot 1.25 ug/ml
NP_006796
heterogeneous nuclear ribonucleoprotein A0, hnRNA binding protein, hnRNPA0, HNRPA0hnRNP A0
Mouse
IgM Kappa
Protein A or G purified
RUO
Primary
10949
Western Blot
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
HNRNPA0
The immunogen for this antibody is HNRNPA0.
31 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit