Learn More
HNRNPL Mouse anti-Human, Clone: OTI9F11, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF805577
Catalog No. 50-166-7487
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Heterogeneous nuclear RNAs which include mRNA precursors and mature mRNAs are associated with specific proteins to form heterogenous ribonucleoprotein complexes. Heterogeneous nuclear ribonucleoprotein L is among the proteins that are stably associated with hnRNP complexes and along with other hnRNP proteins is likely to play a major role in the formation, packaging, processing, and function of mRNA. Heterogeneous nuclear ribonucleoprotein L is present in the nucleoplasm as part of the HNRP complex. HNRP proteins have also been identified outside of the nucleoplasm. Exchange of hnRNP for mRNA-binding proteins accompanies transport of mRNA from the nucleus to the cytoplasm. Since HNRP proteins have been shown to shuttle between the nucleus and the cytoplasm, it is possible that they also have cytoplasmic functions. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
HNRNPL
Monoclonal
Unconjugated
HNRNPL
Lyophilized
HNRNPL
Human recombit protein fragment corresponding to amino acids 270-502 of human HNRNPL produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI9F11
PBS with 8% trehalose and no preservative; pH 7.3
P14866
HNRPL, P/OKcl.14, hnRNP-L
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
3191