Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HNRNPU Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109827
Description
Immunogen sequence: PKDEDYKQRT QKKAEVEGKD LPEHAVLKMK GNFTLPEVAE CFDEITYVEL QKEEAQKLLE QYKEESKKAL PPEKKQNTGS KKSNKNKSGK NQFNRGGGHR GRGGFNMRGG NFRGGAPGNR GGYNRRGNMP QRGGGGGGSG GIGYPYPRAP VFPGRGSYSN RGNYNRGGMP NRGNYNQNFR GRGNNRGYKN QSQGYNQWQQ GQFWGQKPWS QHYHQGYY.
The protein encoded by this gene is a receptor for activated C-kinase (RACK) protein. The encoded protein has been shown to bind in vitro to activated protein kinase C beta I. In addition, this protein is a cutaneous T-cell lymphoma-associated antigen. Finally, the protein contains a bromodomain and two zinc fingers, and is thought to be a transcriptional regulator. Multiple transcript variants encoding several different isoforms have been found for this gene.
Specifications
HNRNPU | |
Polyclonal | |
Unconjugated | |
Hnrnpu | |
AA408410; AI256620; AL024194; AL024437; AW557595; C1orf199; C86794; FLJ30202; FLJ37978; GRIP120; Heterogeneous nuclear ribonucleoprotein U; heterogeneous nuclear ribonucleoprotein U (scaffold attachment factor A); heterogenous nuclear ribonucleoprotein U; hnRNP U; HNRNPU; HNRNPU antisense RNA 1; HNRNPU-AS1; Hnrpu; NCRNA00201; nuclear matrix protein sp120; Nuclear p120 ribonucleoprotein; p120; p120 nuclear protein; pp120; rCG_20317; SAFA; SAF-A; scaffold attachment factor A; Scaffold-attachment factor A; SN1; SP120; system N1 Na+ and H+-coupled glutamine transporter; transporter protein; transporter protein; system N1 Na+ and H+-coupled glutamine transporter; U21.1; U211 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
117280, 3192, 51810 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
1.60 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q00839, Q8VEK3 | |
Hnrnpu | |
Recombinant fusion protein containing a sequence corresponding to amino acids 608-825 of human hnRNP U (NP_1140322). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction