Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HNRPH3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen HNRPH3
Applications Immunofluorescence
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB096057
View Documents
Novus Biologicals
NBP304683100UL
100 μg
Each for $320.71
Add to cart
 
NB096058
View Documents
Novus Biologicals
NBP30468320UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

HNRPH3 Polyclonal antibody specifically detects HNRPH3 in Human samples. It is validated for Immunofluorescence
Specifications

Specifications

HNRPH3
Unconjugated
Rabbit
Human
Heterogeneous nuclear ribonucleoprotein 2H9, heterogeneous nuclear ribonucleoprotein H3, heterogeneous nuclear ribonucleoprotein H3 (2H9), hnRNP 2H9,2H9hnRNP H3, HNRPH3FLJ34092
Recombinant fusion protein containing a sequence corresponding to amino acids 1-331 of human HNRPH3 (NP_067676.2).
Polyclonal
Affinity purified
Immunofluorescence
Purified
RUO
PBS with 50% glycerol, pH7.3.
3189
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit