Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HNRPLL Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen HNRPLL
Applications Immunohistochemistry, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093444
View Documents
Novus Biologicals
NBP304889100UL
100 μg Each for $320.71
Add to cart
 
NB093445
View Documents
Novus Biologicals
NBP30488920UL
20 μg Each for $138.37
Add to cart
 
Description

Description

HNRPLL Polyclonal antibody specifically detects HNRPLL in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

HNRPLL
Unconjugated
Rabbit
Human, Mouse, Rat
heterogeneous nuclear ribonucleoprotein L-like, hnRNPLL, SRRF, stromal RNA regulating factor, Stromal RNA-regulating factor
Recombinant fusion protein containing a sequence corresponding to amino acids 250-350 of human HNRPLL (NP_612403.2).
Polyclonal
Affinity purified
Immunohistochemistry, Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
92906
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit