Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HO-1/HMOX1/HSP32 Mouse anti-Human, Mouse, Rat, Biotin, Clone: 6B8-2F2, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259338B
Description
HO-1/HMOX1/HSP32 Monoclonal specifically detects HO-1/HMOX1/HSP32 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
HO-1/HMOX1/HSP32 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
HMOX1 | |
His-tagged Rat HO-1 | |
100 μL | |
Primary | |
Detects 32kDa. Does not cross-react with HO-2. | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, Immunocytochemistry, Immunofluorescence | |
6B8-2F2 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
bK286B10, EC 1.14.99.3, heat shock protein, 32-kD, heme oxygenase (decycling) 1, HO, HO1, HO-1heme oxygenase 1, HSP32 | |
Mouse | |
Protein G purified | |
Apoptosis, Cellular Markers, ER Markers, Lipid and Metabolism | |
3162 | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title