Learn More
HO-1/HMOX1/HSP32 Mouse anti-Human, Mouse, Rat, DyLight 550, Clone: 6B8-2F2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259338R
Catalog No. NBP259338R
Add to cart
Description
Description
HO-1/HMOX1/HSP32 Monoclonal antibody specifically detects HO-1/HMOX1/HSP32 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
HO-1/HMOX1/HSP32
6B8-2F2
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
HMOX1
His-tagged Rat HO-1
Protein G purified
Apoptosis, Cellular Markers, ER Markers, Lipid and Metabolism
Primary
3162
Human, Mouse, Rat
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
bK286B10, EC 1.14.99.3, heat shock protein, 32-kD, heme oxygenase (decycling) 1, HO, HO1, HO-1heme oxygenase 1, HSP32
Mouse
IgG1 Kappa
100 ul
Store at 4C in the dark.
Monoclonal
Detects 32kDa. Does not cross-react with HO-2.