Learn More
Enzo Life SciencesSupplier Diversity Partner HO-1 monoclonal antibody (HO-1-2) (PE conjugate) (50 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADIOSA111PED
Clone: HO-1-2. Host: Mouse. Isotype: IgG2b. Immunogen: Native rat HO-1. UniProt: P06762. Source: Purified from ascites. Species Reactivity: Human, Mouse, Rat, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog. Applications: Flow Cytometry. Recommended dilutions: Flow Cytometry (1:100). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 0.09% sodium azide.
Catalog No. 50-667-007