Learn More
Enzo Life SciencesSupplier Diversity Partner IMMUNOSET HO-1 (human), ELISA development set (96 wells)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADI960800
The IMMUNOSET HO-1 (human), EIA development set contains sufficient reagents for the development of 5 x 96 well EIA plates which provides analysis of ~200 samples in duplicate. Save time, money, and sample by developing your own EIA kits. Simply coat your plates and go! This kit provides superior accuracy with fully quantitative results compared to Western blot analysis and has low reactivity with related molecules. Alternative name: Heme oxygenase 1. Contents: Capture antibody, Standard, Detection antibody, SA-HRP. Application: ELISA, Colorimetric detection. Assay Time: Plate Coating - Overnight + 1 hour; Assay - 3 hours. Crossreactivity: Specific for human HO-1; does not react with human HO-2 or HO-3. Long Term Storage: +4°C
Catalog No. 50-658-683