Learn More
Enzo Life SciencesSupplier Diversity Partner HO-1 (rat), ELISA kit (1x96wells)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADIEKS810A
Highly sensitive colorimetric immunometric enzyme immunoassay kit with results in < 3 hours. The HO-1 (rat), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 450 nm. Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis. Alternative Name: Heme oxygenase 1. Sensitivity: 0.036 ng/ml (range 0.195 - 12.5 ng/ml). Assay Time: <3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Rat. Contents: Microtiter plate, 5X Extraction reagent 2, Standard, Sample diluent, Wash buffer concentrate, Antibody, Conjugate, TMB Substrate, Stop solution 2
Catalog No. 50-659-138