Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HO-2/HMOX2 Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI1C2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270895AF647

Catalog No. N270895A647


Add to cart

Description

Description

HO-2/HMOX2 Monoclonal antibody specifically detects HO-2/HMOX2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications

Specifications

HO-2/HMOX2
OTI1C2
Western Blot, Flow Cytometry, Immunohistochemistry
Affinity Purified, Conjugated
HMOX2
Full length recombinant protein of human HO-2/HMOX2 (NP_002125) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
heme oxygenase (decycling) 2, heme oxygenase 2, HO2, HO-2EC 1.14.99.3
Mouse
IgG1
0.1 mL
Primary
3163.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit