Learn More
Hoefer™ Combs for HE 99X Max Submarine Electrophoresis Unit
Backless combs for use with Hoefer HE99X Max Submarine Electrophoresis Unit
Manufacturer:  Hoefer™ HE91A203.0
 View more versions of this product
Catalog No. 03-500-142
  Check Availability
Add to cart
Requires : Comb back must be ordered separately (see ordering table)
Specifications Description & Specifications
Specifications
0.11 in.
0.19 in.
Backless Comb
20
70.5μL
0.3cm
0.5cm
HE99X max submarine electrophoresis unit
0.18 in.
0.47cm