Learn More
Hoefer™ Combs for SE 250, SE 260, or miniVE Vertical Electrophoresis Units
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
Supplier: Hoefer™ SE211AR1.0
Description
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
- Lightweight
- Virtually unbreakable
Specifications
SE 250, SE 260, MiniVE™ Vertical Electrophoresis System | |
PTFE | |
0.03 in. | |
68μL/ mm | |
6.8 cm | |
1.3 cm |
Comb | |
1 Prep, 1 Reference | |
0.1 cm | |
2.6 in. | |
0.51 in. |